Learn More
Abcam Human MRPS7 knockout HeLa cell line
Descrizione
MRPS7 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 11 bp deletion in exon 1 and 2 bp deletion in exon 1 and 31 bp deletion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Specifica
| Quantità | 2 x 1,000,000 Cells |
| Status giuridico | RUO |