Learn More
Abnova™ Human MS4A6E Full-length ORF (NP_640342.1, 1 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00245802-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the membrane-spanning 4A gene family. Members of this nascent protein family are characterized by common structural features and similar intron/exon splice boundaries and display unique expression patterns among hematopoietic cells and nonlymphoid tissues. The gene encoding this protein is localized to 11q12.3, among a cluster of family members. [provided by RefSeq]
Sequence: MTSQPISNETIIMLPSNVINFSQAEKPEPTNQGQDSLKKRLQAKVKVIGVHSSLAGSILSALSALVGFILLSVNPAALNPASLQCKLDEKDIPTRLLLSYDYHSPYTMDCHRAKASLAGTLSLMLVSTVLEFCLAVLTAVLQWKQTVSpecifica
NP_640342.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
42.3kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC126832 | |
MS4A6E | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
245802 | |
MS4A6E (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MTSQPISNETIIMLPSNVINFSQAEKPEPTNQGQDSLKKRLQAKVKVIGVHSSLAGSILSALSALVGFILLSVNPAALNPASLQCKLDEKDIPTRLLLSYDYHSPYTMDCHRAKASLAGTLSLMLVSTVLEFCLAVLTAVLQWKQTV | |
RUO | |
MS4A6E | |
Wheat Germ (in vitro) | |
GST | |
Liquid |