Learn More
Abnova™ Human MYO1E Partial ORF (NP_004989.2, 918 a.a. - 1014 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00004643-Q01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16131605
  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile
Descrizione
Descrizione
Sequence: VSIGPGLPKNSRPTRRNTTQNTGYSSGTQNANYPVRAAPPPPGYHQNGVIRNQYVPYPHAPGSQRSNQKSLYTSMARPPLPRQQSTSSDRVSQTPES
Specifica
Specifica
NP_004989.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.41kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
VSIGPGLPKNSRPTRRNTTQNTGYSSGTQNANYPVRAAPPPPGYHQNGVIRNQYVPYPHAPGSQRSNQKSLYTSMARPPLPRQQSTSSDRVSQTPES
RUO
MYO1E
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4643
MYO1E (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HuncM-IC/MGC104638/MYO1C
MYO1E
Recombinant
wheat germ expression system