missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NAP1L4 Partial ORF (NP_005960, 1 a.a. - 82 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00004676-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16171655

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the nucleosome assembly protein (NAP) family which can interact with both core and linker histones. It can shuttle between the cytoplasm and nucleus, suggesting a role as a histone chaperone. This gene is one of several located near the imprinted gene domain of 11p15.5, an important tumor-suppressor gene region. Alterations in this region have been associated with the Beckwith-Wiedemann syndrome, Wilms tumor, rhabdomyosarcoma, adrenocortical carcinoma, and lung, ovarian, and breast cancer. [provided by RefSeq]

Sequence: MADHSFSDGVPSDSVEAAKNASNTEKLTDQVMQNPRVLAALQERLDNVPHTPSSYIETLPKAVKRRINALKQLQVRCAHIEA
Specifica

Specifica

NP_005960
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.76kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MADHSFSDGVPSDSVEAAKNASNTEKLTDQVMQNPRVLAALQERLDNVPHTPSSYIETLPKAVKRRINALKQLQVRCAHIEA
RUO
NAP1L4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
4676
NAP1L4 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC4565/NAP2/NAP2L/hNAP2
NAP1L4
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali