missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human NDUFS7 Full-length ORF (NP_077718.2, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00374291-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16160354

  • 302.00€ / 10µg


Spedizione stimata
27-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MAVLSAPGLRGFRILGLRSSVGLAVQARGVHQSVATDGPSSTQPALPKARAVAPKPSSRGEYVVAKLDDLVNWARRSSLWPMTFGLACCAVEMMHMAAPRYDMDRFGVVFRASPRQSDVMIVAGTLTNKMAPALRKVYDQMPEPRYVVSMGSCANGGGYYHYSYSVVRGCDRIVPVDIYIPGCPPTAEALLYGILQLQRKIKRERRLQIWYRR
Specifica

Specifica

NP_077718.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
50kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CI-20KD|FLJ45860|FLJ46880|MGC120002|MY017|PSST
NDUFS7
Yes
wheat germ expression system
Liquid
374291
NDUFS7 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAVLSAPGLRGFRILGLRSSVGLAVQARGVHQSVATDGPSSTQPALPKARAVAPKPSSRGEYVVAKLDDLVNWARRSSLWPMTFGLACCAVEMMHMAAPRYDMDRFGVVFRASPRQSDVMIVAGTLTNKMAPALRKVYDQMPEPRYVVSMGSCANGGGYYHYSYSVVRGCDRIVPVDIYIPGCPPTAEALLYGILQLQRKIKRERRLQIWYRR
RUO
NDUFS7
Wheat Germ (in vitro)
GST
Videos
SDS
Documenti