missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ Human Nidogen Recombinant Protein, PeproTech®

Codice prodotto. 30019982
Cambia vista
Click to view available options
Quantità:
1 mg
100 μg
25 μg
250 μg
5 μg
500 μg
Dimensione della confezione:
100µg
1mg
250µg
25µg
500µg
5µg
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
30019982 1 mg 1mg
30019789 5 μg 5µg
30019358 500 μg 500µg
30019830 25 μg 25µg
30019712 250 μg 250µg
30019473 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 30019982 Fornitore Gibco™ N. del fornitore 140171MG

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Recombinant Protein

This gene encodes a member of the nidogen family of basement membrane glycoproteins. The protein interacts with several other components of basement membranes, and may play a role in cell interactions with the extracellular matrix.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso P14543
Da utilizzare con (applicazione) Functional Assay
Formato Lyophilized
Formulazione protein with no preservative
ID gene (immissione) 4811
Peso molecolare 133.5 kDa
Nome Human Nidogen
Quantità 1 mg
Status giuridico RUO
Concentrazione di endotossine <1 EU/ μg
Alias gene A630025O17; enactin; Ent; Entactin; entactin 1; entactin-1; Nid; NID 1; NID1; NID-1; Nidogen (enactin); nidogen (entactin); nidogen 1; Nidogen precursor (Entactin); nidogen-1; Nidogen-1;
Nome comune Nidogen
Attività biologica Determined by the ability of the immobilized protein to support the adhesion of HUVEC cells.
Coniugato Unconjugated
Recombinant Recombinant
Sequenza LSRQELFPFG PGQGDLELED GDDFVSPALE LSGALRFYDR SDIDAVYVTT NGIIATSEPP AKESHPGLFP PTFGAVAPFL ADLDTTDGLG KVYYREDLSP SITQRAAECV HRGFPEISFQ PSSAVVVTWE SVAPYQGPSR DPDQKGKRNT FQAVLASSDS SSYAIFLYPE DGLQFHTTFS KKENNQVPAV VAFSQGSVGF LWKSNGAYNI FANDRESVEN LAKSSNSGQQ GVWVFEIGSP ATTNGVVPAD VILGTEDGAE YDDEDEDYDL ATTRLGLEDV GTTPFSYKAL RRGGADTYSV PSVLSPRRAA TERPLGPPTE RTRSFQLAVE TFHQQHPQVI DVDEVEETGV VFSYNTDSRQ TCANNRHQCS VHAECRDYAT GFCCSCVAGY TGNGRQCVAE GSPQRVNGKV KGRIFVGSSQ VPIVFENTDL HSYVVMNHGR SYTAISTIPE TVGYSLLPLA PVGGIIGWMF AVEQDGFKNG FSITGGEFTR QAEVTFVGHP GNLVIKQRFS GIDEHGHLTI DTELEGRVPQ IPFGSSVHIE PYTELYHYST SVITSSSTRE YTVTEPERDG ASPSRIYTYQ WRQTITFQEC VHDDSRPALP STQQLSVDSV FVLYNQEEKI LRYALSNSIG PVREGSPDAL QNPCYIGTHG CDTNAACRPG PRTQFTCECS IGFRGDGRTC YDIDECSEQP SVCGSHTICN NHPGTFRCEC VEGYQFSDEG TCVAVVDQRP INYCETGLHN CDIPQRAQCI YTGGSSYTCS CLPGFSGDGQ ACQDVDECQP SRCHPDAFCY NTPGSFTCQC KPGYQGDGFR CVPGEVEKTR CQHEREHILG AAGATDPQRP IPPGLFVPEC DAHGHYAPTQ CHGSTGYCWC VDRDGREVEG TRTRPGMTPP CLSTVAPPIH QGPAVPTAVI PLPPGTHLLF AQTGKIERLP LEGNTMRKTE AKAFLHVPAK VIIGLAFDCV DKMVYWTDIT EPSIGRASLH GGEPTTIIRQ DLGSPEGIAV DHLGRNIFWT DSNLDRIEVA KLDGTQRRVL FETDLVNPRG IVTDSVRGNL YWTDWNRDNP KIETSYMDGT NRRILVQDDL GLPNGLTFDA FSSQLCWVDA GTNRAECLNP SQPSRRKALE GLQYPFAVTS YGKNLYFTDW KMNSVVALDL AISKETDAFQ PHKQTRLYGI TTALSQCPQG HNYCSVNNGG CTHLCLATPG SRTCRCPDNT LGVDCIEQK
Content And Storage -20°C
Sistema di espressione CHO cells
Forma Lyophilized
Purity or Quality Grade ≥ 95% by SDS-PAGE gel and HPLC analyses.
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.