missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human P11 Partial ORF (NP_006016.1, 270 a.a. - 369 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008909-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16129635

  • 528.00€ / 25µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a protein with protease activity. Its specific function has not been determined. The protein may be useful as a tumor marker. [provided by RefSeq]

Sequence: EGLVDYYSHIYDGPWDSYPDVLAMQFNWDGYYKEVGSAFIGSSPEFEFALYSLCFIARPGKVCQLSLGGYPLAVRTYTWDKSTYGNGKKYIATAYIVSST
Specifica

Specifica

NP_006016.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EGLVDYYSHIYDGPWDSYPDVLAMQFNWDGYYKEVGSAFIGSSPEFEFALYSLCFIARPGKVCQLSLGGYPLAVRTYTWDKSTYGNGKKYIATAYIVSST
RUO
P11
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
8909
P11 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC133268/PP11/PRSS26
P11
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali