missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human p38 beta/MAPK11 (NP_002410.1) VersaClone cDNA
Specifications
Specifications
| Gene | p38 beta/MAPK11 |
| Numero di accesso | NP_002410.1 |
| ID gene (immissione) | 5600 |
| Alias gene | EC 2.7.11, EC 2.7.11.24, MAP kinase 11, MAP kinase p38 beta, MAPK 11, MAPK11, mitogen-activated protein kinase 11, Mitogen-activated protein kinase p38 beta, mitogen-activated protein kinase p38-2, MPK11, p38b, p38Beta, P38BETA2, PRKM11, SAPK2B, SAPK2p38-2P38B, Stress-activated protein kinase 2, stress-activated protein kinase-2, stress-activated protein kinase-2b |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?