Learn More
Abnova™ Human PAPSS2 Full-length ORF (NP_004661.2, 1 a.a. - 614 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifica
Numero di accesso | NP_004661.2 |
---|---|
Da utilizzare con (applicazione) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulazione | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID gene (immissione) | 9060 |
Peso molecolare | 95.9kDa |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16173232
|
Abnova™
H00009060-P01.25UG |
25 ug |
508.00€
25µg |
Spedizione stimata: 05-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16163232
|
Abnova™
H00009060-P01.10UG |
10 ug |
335.00€
10µg |
Spedizione stimata: 05-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Sulfation is a common modification of endogenous (lipids, proteins, and carbohydrates) and exogenous (xenobiotics and drugs) compounds. In mammals, the sulfate source is 3'-phosphoadenosine 5'-phosphosulfate (PAPS), created from ATP and inorganic sulfate. Two different tissue isoforms encoded by different genes synthesize PAPS. This gene encodes one of the two PAPS synthetases. Defects in this gene cause the Pakistani type of spondyloepimetaphyseal dysplasia. Two alternatively spliced transcript variants that encode different isoforms have been described for this gene. [provided by RefSeq]
Sequence: MSGIKKQKTENQQKSTNVVYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTISFALEEYLVSHAIPCYSLDGDNVRHGLNRNLGFSPGDREENIRRIAEVAKLFADAGLVCITSFISPFAKDRENARKIHESAGLPFFEIFVDAPLNICESRDVKGLYKRARAGEIKGFTGIDSDYEKPETPERVLKTNLSTVSDCVHQVVELLQEQNIVPYTIIKDIHELFVPENKLDHVRAEAETLPSLSITKLDLQWVQVLSEGWATPLKGFMREKEYLQVMHFDTLLDDGVINMSIPIVLPVSAEDKTRLEGCSKFVLAHGGRRVAILRDAEFYEHRKEERCSRVWGTTCTKHPHIKMVMESGDWLVGGDLQVLEKIRWNDGLDQYRLTPLELKQKCKEMNADAVFAFQLRNPVHNGHALLMQDTRRRLLERGYKHPVLLLHPLGGWTKDDDVPLDWRMKQHAAVLEEGVLDPKSTIVAIFPSPMLYAGPTEVQWHCRSRMIAGANFYIVGRDPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLEKNSpecifica
NP_004661.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
95.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MSGIKKQKTENQQKSTNVVYQAHHVSRNKRGQVVGTRGGFRGCTVWLTGLSGAGKTTISFALEEYLVSHAIPCYSLDGDNVRHGLNRNLGFSPGDREENIRRIAEVAKLFADAGLVCITSFISPFAKDRENARKIHESAGLPFFEIFVDAPLNICESRDVKGLYKRARAGEIKGFTGIDSDYEKPETPERVLKTNLSTVSDCVHQVVELLQEQNIVPYTIIKDIHELFVPENKLDHVRAEAETLPSLSITKLDLQWVQVLSEGWATPLKGFMREKEYLQVMHFDTLLDDGVINMSIPIVLPVSAEDKTRLEGCSKFVLAHGGRRVAILRDAEFYEHRKEERCSRVWGTTCTKHPHIKMVMESGDWLVGGDLQVLEKIRWNDGLDQYRLTPLELKQKCKEMNADAVFAFQLRNPVHNGHALLMQDTRRRLLERGYKHPVLLLHPLGGWTKDDDVPLDWRMKQHAAVLEEGVLDPKSTIVAIFPSPMLYAGPTEVQWHCRSRMIAGANFYIVGRDPAGMPHPETKKDLYEPTHGGKVLSMAPGLTSVEIIPFRVAAYNKAKKAMDFYDPARHNEFDFISGTRMRKLAREGENPPDGFMAPKAWKVLTDYYRSLEKN | |
RUO | |
PAPSS2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9060 | |
PAPSS2 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ATPSK2/SK2 | |
PAPSS2 | |
Recombinant | |
wheat germ expression system |