Learn More
Abnova™ Human PCBP4 Partial ORF (NP_127503, 130 a.a. - 239 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057060-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the KH-domain protein subfamily. Proteins of this subfamily, also referred to as alpha-CPs, bind to RNA with a specificity for C-rich pyrimidine regions. Alpha-CPs play important roles in post-transcriptional activities and have different cellular distributions. This gene is induced by the p53 tumor suppressor, and the encoded protein can suppress cell proliferation by inducing apoptosis and cell cycle arrest in G(2)-M. This gene's protein is found in the cytoplasm, yet it lacks the nuclear localization signals found in other subfamily members. Multiple alternatively spliced transcript variants have been described, but the full-length nature for only some has been determined. [provided by RefSeq]
Sequence: TTGAQVQVAGDLLPNSTERAVTVSGVPDAIILCVRQICAVILESPPKGATIPYHPSLSLGTVLLSANQGFSVQGQYGAVTPAEVTKLQQLSSHAVPFATPSVVPGLDPGTSpecifica
NP_127503 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.62kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TTGAQVQVAGDLLPNSTERAVTVSGVPDAIILCVRQICAVILESPPKGATIPYHPSLSLGTVLLSANQGFSVQGQYGAVTPAEVTKLQQLSSHAVPFATPSVVPGLDPGT | |
RUO | |
PCBP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
57060 | |
PCBP4 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
LIP4/MCG10 | |
PCBP4 | |
Recombinant | |
wheat germ expression system |