Learn More
ACROBiosystems Human PD-1 / PDCD1 Full Length Protein, His Tag
718.53€ - 6532.14€
Specifica
Numero di accesso | NP_005009.2 |
---|---|
Peso molecolare | 30.7 kDa |
Sorgente | Human PD-1 Full Length, His Tag (PD1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Leu 288 (Accession # Q15116-1). |
Immunogeno | Leu 25 - Leu 288 |
Requisiti di stoccaggio | -20°C |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
17226410
|
ACROBiosystems
PD1-H52H6-20UG |
20 μg |
718.53€
20µg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
17236410
|
ACROBiosystems
PD1-H52H6-500UG |
500 μg |
6532.14€
500µg |
Spedizione stimata: 30-04-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Programmed cell death protein 1 (PD-1) is also known as CD279 and PDCD1, is a type I membrane protein and is a member of the extended CD28/CTLA-4 family of T cell regulators. PDCD1 is expressed on the surface of activated T cells, B cells, macrophages, myeloid cells and a subset of thymocytes. PD-1 has two ligands, PD-L1 and PD-L2, which are members of the B7 family. PD-L1 is expressed on almost all murine tumor cell lines, including PA1 myeloma, P815 mastocytoma, and B16 melanoma upon treatment with IFN-γ. PD-L2 expression is more restricted and is expressed mainly by DCs and a few tumor lines. PD1 inhibits the T-cell proliferation and production of related cytokines including IL-1, IL-4, IL-10 and IFN-γ by suppressing the activation and transduction of PI3K/AKT pathway. In addition, coligation of PD1 inhibits BCR-mediating signal by dephosphorylating key signal transducer. In vitro, treatment of anti-CD3 stimulated T cells with PD-L1-Ig results in reduced T cell proliferation and IFN-γ secretion. Monoclonal antibodies targeting PD-1 that boost the immune system are being developed for the treatment of cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 30.7 kDa. The protein migrates as 40-120 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifica
NP_005009.2 | |
Human PD-1 Full Length, His Tag (PD1-H52H6) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Leu 288 (Accession # Q15116-1). | |
-20°C | |
PD-1 | |
Human | |
50 mM Tris, 100 mM NaCl, pH8.0 | |
HEK293 | |
Immune Checkpoint Proteins |
30.7 kDa | |
Leu 25 - Leu 288 | |
PDCD1,PD1,CD279,SLEB2 | |
Unconjugated | |
His Tag | |
Less than 1.0 EU / μg by the LAL method. | |
Powder | |
>80% as determined by SDS-PAGE. |