Learn More
Abnova™ Human PEX16 Partial ORF (NP_004804.1, 1 a.a. - 100 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00009409-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
The protein encoded by this gene is an integral peroxisomal membrane protein. An inactivating nonsense mutation localized to this gene was observed in a patient with Zellweger syndrome of the complementation group CGD/CG9. Expression of this gene product morphologically and biochemically restores the formation of new peroxisomes, suggesting a role in peroxisome organization and biogenesis. Alternative splicing has been observed for this gene and two variants have been described. [provided by RefSeq]
Sequence: MEKLRLLGLRYQEYVTRHPAATAQLETAVRGFSYLLAGRFADSHELSELVYSASNLLVLLNDGILRKELRKKLPVSLSQQKLLTWLSVLECVEVFMEMGASpecifica
NP_004804.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.74kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MEKLRLLGLRYQEYVTRHPAATAQLETAVRGFSYLLAGRFADSHELSELVYSASNLLVLLNDGILRKELRKKLPVSLSQQKLLTWLSVLECVEVFMEMGA | |
RUO | |
PEX16 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
9409 | |
PEX16 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PEX16 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |