Learn More
Human Phospho-RelA/NFkB p65 (S536) Antibody, R&D Systems™
GREENER_CHOICE
Codice prodotto. 18029981
Cambia vista
Click to view available options
Quantità:
100 μg
Dimensione della confezione:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità unitSize
18029981 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 18029981 Fornitore R&D Systems N. del fornitore MAB7226
Mouse Monoclonal Antibody
RelA/NFkB p65 Monoclonal specifically detects RelA/NFkB p65 in Human samples. It is validated for Western Blot, Immunocytochemistry.
TRUSTED_SUSTAINABILITY
Specifica
Antigene RelA/NFkB p65 (p ser536)
Applicazioni Western Blot, Immunocytochemistry
Classificazione Monoclonal
Clone 817403
Concentrazione LYOPH
Coniugato Unconjugated
Diluizione Western Blot 2 ug/mL, Immunocytochemistry 8-25 ug/mL
Formulazione Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Alias gene MGC131774, nf kb p65, NFkB p65, NF-kB p65, NFKB3, NFKB3v-rel avian reticuloendotheliosis viral oncogene homolog A (nuclear factor ofkappa light polypeptide gene enhancer in B-cells 3 (p65)), Nuclear factor NF-kappa-B p65 subunit, Nuclear factor of kappa light polypeptide gene enhancer in B-cells 3transcription factor p65, p65, p65RelA, RelA, rela p65, v-rel reticuloendotheliosis viral oncogene homolog A (avian), v-rel reticuloendotheliosis viral oncogene homolog A, nuclear factor of kappalight polypeptide gene enhancer in B-cells 3, p65
Simboli geni RELA
Specie ospite Mouse
Immunogeno Phosphopeptide containing the RelA/NF kappa B p65 S536 site
Metodo di purificazione Protein A or G purified from hybridoma culture supernatant
Quantità 100 μg
Status giuridico RUO
Primario o secondario Primary
ID gene (immissione) 5970
Test di specificità Detects human RelA/NF kappa B p65 when phosphorylated at S536.
Ricostituzione Sterile PBS to a final concentration of 0.5 mg/mL.
Target Species Human
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Forma Purified
Isotype IgG1