missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PLCD3 Partial ORF (NP_588614, 514 a.a. - 613 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00113026-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16161357

  • 335.00€ / 10µg
Spedizione stimata: 10-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a member of the phospholipase C family, which catalyze the hydrolysis of phosphatidylinositol 4,5-bisphosphate to generate the second messengers diacylglycerol and inositol 1,4,5-trisphosphate (IP3). Diacylglycerol and IP3 mediate a variety of cellular responses to extracellular stimuli by inducing protein kinase C and increasing cytosolic Ca(2+) concentrations. This enzyme localizes to the plasma membrane and requires calcium for activation. Its activity is inhibited by spermine, sphingosine, and several phospholipids. [provided by RefSeq]

Sequence: AAAQRRLAKQISPELSALAVYCHATRLRTLHPAPNAPQPCQVSSLSERKAKKLIREAGNSFVRHNARQLTRVYPLGLRMNSANYSPQEMWNSGCQLVALN
Specifica

Specifica

NP_588614
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
AAAQRRLAKQISPELSALAVYCHATRLRTLHPAPNAPQPCQVSSLSERKAKKLIREAGNSFVRHNARQLTRVYPLGLRMNSANYSPQEMWNSGCQLVALN
RUO
PLCD3
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
113026
PLCD3 (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC71172
PLCD3
Recombinant
wheat germ expression system
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali