missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PODXL2 Full-length ORF (NP_056535.1, 1 a.a. - 605 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00050512-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16159392

  • 335.00€ / 10µg
Spedizione stimata: 27-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

Sequence: MGRLLRAARLPPLLSPLLLLLVGGAFLGACVAGSDEPGPEGLTSTSLLDLLLPTGLEPLDSEEPSETMGLGAGLGAPGSGFPSEENEESRILQPPQYFWEEEEELNDSSLDLGPTADYVFPDLTEKAGSIEDTSQAQELPNLPSPLPKMNLVEPPWHMPPREEEEEEEEEEEREKEEVEKQEEEEEEELLPVNGSQEEAKPQVRDFSLTSSSQTPGATKSRHEDSGDQASSGVEVESSMGPSLLLPSVTPTTVTPGDQDSTSQEAEATVLPAAGLGVEFEAPQEASEEATAGAAGLSGQHEEVPALPSFPQTTAPSGAEHPDEDPLGSRTSASSPLAPGDMELTPSSATLGQEDLNQQLLEGQAAEAQSRIPWDSTQVICKDWSNLAGKNYIILNMTENIDCEVFRQHRGPQLLALVEEVLPRHGSGHHGAWHISLSKPSEKEQHLLMTLVGEQGVVPTQDVLSMLGDIRRSLEEIGIQNYSTTSSCQARASQVRSDYGTLFVVLVVIGAICIIIIALGLLYNCWQRRLPKLKHVSHGEELRFVENGCHDNPTLDVASDSQSEMQEKHPSLNGGGALNGPGSWGALMGGKRDPEDSDVFEEDTHL
Specifica

Specifica

NP_056535.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
91.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
PODLX2
PODXL2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
50512
PODXL2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MGRLLRAARLPPLLSPLLLLLVGGAFLGACVAGSDEPGPEGLTSTSLLDLLLPTGLEPLDSEEPSETMGLGAGLGAPGSGFPSEENEESRILQPPQYFWEEEEELNDSSLDLGPTADYVFPDLTEKAGSIEDTSQAQELPNLPSPLPKMNLVEPPWHMPPREEEEEEEEEEEREKEEVEKQEEEEEEELLPVNGSQEEAKPQVRDFSLTSSSQTPGATKSRHEDSGDQASSGVEVESSMGPSLLLPSVTPTTVTPGDQDSTSQEAEATVLPAAGLGVEFEAPQEASEEATAGAAGLSGQHEEVPALPSFPQTTAPSGAEHPDEDPLGSRTSASSPLAPGDMELTPSSATLGQEDLNQQLLEGQAAEAQSRIPWDSTQVICKDWSNLAGKNYIILNMTENIDCEVFRQHRGPQLLALVEEVLPRHGSGHHGAWHISLSKPSEKEQHLLMTLVGEQGVVPTQDVLSMLGDIRRSLEEIGIQNYSTTSSCQARASQVRSDYGTLFVVLVVIGAICIIIIALGLLYNCWQRRLPKLKHVSHGEELRFVENGCHDNPTLDVASDSQSEMQEKHPSLNGGGALNGPGSWGALMGGKRDPEDSDVFEEDTHL
RUO
PODXL2
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali