missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPBP Partial ORF (AAH28217, 37 a.a. - 128 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005473-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16159391

  • 458.00€ / 25µg


Spedizione stimata
24-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: TKGQTKRNLAKGKEESLDSDLYAELRCMCIKTTSGIHPKNIQSLEVIGKGTHCNQVEVIATLKDGRKICLDPDAPRIKKIVQKKLAGDESAD
Specifica

Specifica

AAH28217
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
35.75kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
TKGQTKRNLAKGKEESLDSDLYAELRCMCIKTTSGIHPKNIQSLEVIGKGTHCNQVEVIATLKDGRKICLDPDAPRIKKIVQKKLAGDESAD
RUO
PPBP
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
5473
PPBP (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
B-TG1|Beta-TG|CTAP-III|CTAP3|CTAPIII|CXCL7|LA-PF4|LDGF|MDGF|NAP-2|PBP|SCYB7|TC1|TC2|TGB|TGB1|THBGB|THBGB1
PPBP
Yes
Videos
Documenti