missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPM1G Partial ORF (NP_817092, 462 a.a. - 544 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00005496-Q01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16159411

  • 302.00€ / 10µg


Spedizione stimata
18-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: QRDENGELRLLSSIVEELLDQCLAPDTSGDGTGCDNMTCIIICFKPRNTAELQPESGKRKLEEVLSTEGAEENGNSDKKKKAK
Specifica

Specifica

NP_817092
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
34.87kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
QRDENGELRLLSSIVEELLDQCLAPDTSGDGTGCDNMTCIIICFKPRNTAELQPESGKRKLEEVLSTEGAEENGNSDKKKKAK
RUO
PPM1G
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
5496
PPM1G (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC1675|MGC2870|PP2CG|PP2CGAMMA|PPP2CG
PPM1G
Yes
Videos
Documenti