missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PPP2R2B Partial ORF (AAH31790, 101 a.a. - 200 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifica

Numero di accesso AAH31790
Da utilizzare con (applicazione) Antibody Production, ELISA, Protein Array, Western Blot
Formulazione 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID gene (immissione) 5521
Peso molecolare 36.63kDa
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
16149451
Vedere i documenti Dettagli della promozione
Abnova™
H00005521-Q01.10UG
10 ug
335.00€
10µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 27-05-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16159451
Vedere i documenti Dettagli della promozione
Abnova™
H00005521-Q01.25UG
25 ug
508.00€
25µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 27-05-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

The product of this gene belongs to the phosphatase 2 regulatory subunit B family. Protein phosphatase 2 is one of the four major Ser/Thr phosphatases, and it is implicated in the negative control of cell growth and division. It consists of a common heteromeric core enzyme, which is composed of a catalytic subunit and a constant regulatory subunit, that associates with a variety of regulatory subunits. The B regulatory subunit might modulate substrate selectivity and catalytic activity. This gene encodes a beta isoform of the regulatory subunit B55 subfamily. Defects in this gene cause autosomal dominant spinocerebellar ataxia 12 (SCA12), a disease caused by degeneration of the cerebellum, sometimes involving the brainstem and spinal cord, and in resulting in poor coordination of speech and body movements. Multiple alternatively spliced variants, which encode different isoforms, have been identified for this gene. The 5' UTR of some of these variants includes a CAG trinucleotide repeat sequence (7-28 copies) that can be expanded to 66-78 copies in cases of SCA12. [provided by RefSeq]

Sequence: QNAAYFLLSTNDKTVKLWKVSERDKRPEGYNLKDEEGRLRDPATITTLRVPVLRPMDLMVEATPRRVFANAHTYHINSISVNSDYETYMSADDLRINLWN
Specifica

Specifica

AAH31790
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
B55-BETA/FLJ95686/MGC24888/PP2A-B55BETA/PP2A-PR55B/PP2AB-BETA/PP2APR55-BETA/PR2AB-BETA/PR2AB55-BETA/PR2APR55-BETA/PR52B/PR55-BETA/SCA12
PPP2R2B
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
5521
PPP2R2B (Human) Recombinant Protein (Q01)
QNAAYFLLSTNDKTVKLWKVSERDKRPEGYNLKDEEGRLRDPATITTLRVPVLRPMDLMVEATPRRVFANAHTYHINSISVNSDYETYMSADDLRINLWN
RUO
PPP2R2B
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Product Certifications