missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human PRKG1 Partial ORF (NP_006249, 73 a.a. - 170 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005592-Q01.25ug
Dettagli aggiuntivi : Peso : 0.02000kg
Descrizione
Sequence: RTKRQAISAEPTAFDIQDLSHVTLPFYPKSPQSKDLIKEAILDNDFMKNLELSQIQEIVDCMYPVEYGKDSCIIKEGDVGSLVYVMEDGKVEVTKEGVSpecifica
NP_006249 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.52kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RTKRQAISAEPTAFDIQDLSHVTLPFYPKSPQSKDLIKEAILDNDFMKNLELSQIQEIVDCMYPVEYGKDSCIIKEGDVGSLVYVMEDGKVEVTKEGV | |
RUO | |
PRKG1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
5592 | |
PRKG1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CGKI/DKFZp686K042/FLJ36117/MGC71944/PGK/PKG/PRKG1B/PRKGR1B/cGKI-BETA/cGKI-alpha | |
PRKG1 | |
Recombinant | |
wheat germ expression system |