Learn More
Abcam Human PTGR1 knockout HeLa cell line
Product Code. 32152637 Shop All Abcam Products
Change view
Click to view available options
Quantità:
2 x 1,000,000 Cells
Unit Size:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantità Unit Size
32152637 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 32152637 Supplier Abcam Supplier No. ab2651692x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 07-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
This item is not returnable. View return policy
PTGR1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 2 bp deletion in exon 2 and Insertion of the selection cassette in exon 2. To order both knockout and wild-type control cells: select 2 x 10000...
PTGR1 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 2 bp deletion in exon 2 and Insertion of the selection cassette in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantità 2 x 1,000,000 Cells
Status giuridico RUO
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.