missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human PTP4A2 Full-length ORF (NP_536316.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008073-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16172262

  • 349.00€ / 10µg
Spedizione stimata: 26-10-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene belongs to a small class of the protein tyrosine phosphatase (PTP) family. PTPs are cell signaling molecules that play regulatory roles in a variety of cellular processes. PTPs in this class contain a protein tyrosine phosphatase catalytic domain and a characteristic C-terminal prenylation motif. This PTP has been shown to primarily associate with plasmic and endosomal membrane through its C-terminal prenylation. This PTP was found to interact with the beta-subunit of Rab geranylgeranyltransferase II (beta GGT II), and thus may function as a regulator of GGT II activity. Overexpression of this gene in mammalian cells conferred a transformed phenotype, which suggested its role in tumorigenesis. Alternatively spliced transcript variants that encode two distinct isoforms have been described. [provided by RefSeq]

Sequence: MNRPAPVEISYENMRFLITHNPTNATLNKFTEELKKYGVTTLVRVCDATYDKAPVEKEGIHVLDWPFDDGAPPPNQIVDDWLNLLKTKFREEPGCCVAVHCVAGLGRAPVLVALALIECGMKYEDAVQFIRQKRRGAFNSKQLLYLEKYRPKMRLRFRDTNGHCCVQ
Specifica

Specifica

NP_536316.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
45.5kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HH13/HH7-2/HU-PP-1/OV-1/PRL-2/PRL2/PTP4A/PTPCAAX2/ptp-IV1a/ptp-IV1b
PTP4A2
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
8073
PTP4A2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNRPAPVEISYENMRFLITHNPTNATLNKFTEELKKYGVTTLVRVCDATYDKAPVEKEGIHVLDWPFDDGAPPPNQIVDDWLNLLKTKFREEPGCCVAVHCVAGLGRAPVLVALALIECGMKYEDAVQFIRQKRRGAFNSKQLLYLEKYRPKMRLRFRDTNGHCCVQ
RUO
PTP4A2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali