Learn More
Abnova™ Human RAB4B Full-length ORF (NP_057238.3, 1 a.a. - 213 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00053916-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
RAB proteins, such as RAB4B, are members of the RAS superfamily of small GTPases that are involved in vesicular trafficking (He et al., 2002 [PubMed 12450215]).[supplied by OMIM]
Sequence: MAETYDFLFKFLVIGSAGTGKSCLLHQFIENKFKQDSNHTIGVEFGSRVVNVGGKTVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNSLAAWLTDARTLASPNIVVILCGNKKDLDPEREVTFLEASRFAQENELMFLETSALTGENVEEAFLKCARTILNKIDSGELDPERMGSGIQYGDASLRQLRQPRSAQAVAPQPCGCSpecifica
NP_057238.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
50kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ78649/MGC52123 | |
RAB4B | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
53916 | |
RAB4B (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAETYDFLFKFLVIGSAGTGKSCLLHQFIENKFKQDSNHTIGVEFGSRVVNVGGKTVKLQIWDTAGQERFRSVTRSYYRGAAGALLVYDITSRETYNSLAAWLTDARTLASPNIVVILCGNKKDLDPEREVTFLEASRFAQENELMFLETSALTGENVEEAFLKCARTILNKIDSGELDPERMGSGIQYGDASLRQLRQPRSAQAVAPQPCGC | |
RUO | |
RAB4B | |
Wheat Germ (in vitro) | |
GST | |
Liquid |