Learn More
Abnova™ Human RANGAP1 Full-length ORF (AAH14044, 1 a.a. - 587 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00005905-P01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
RanGAP1, is a homodimeric 65-kD polypeptide that specifically induces the GTPase activity of RAN, but not of RAS by over 1,000-fold. RanGAP1 is the immediate antagonist of RCC1, a regulator molecule that keeps RAN in the active, GTP-bound state. The RANGAP1 gene encodes a 587-amino acid polypeptide. The sequence is unrelated to that of GTPase activators for other RAS-related proteins, but is 88% identical to Fug1, the murine homolog of yeast Rna1p. RanGAP1 and RCC1 control RAN-dependent transport between the nucleus and cytoplasm. RanGAP1 is a key regulator of the RAN GTP/GDP cycle. [provided by RefSeq]
Sequence: MASEDIAKLAETLAKTQVAGGQLSFKGKSLKLNTAEDAKDVIKEIEDFDSLEALRLEGNTVGVEAARVIAKALEKKSELKRCHWSDMFTGRLRTEIPPALISLGEGLITAGAQLVELDLSDNAFGPDGVQGFEALLKSSACFTLQELKLNNCGMGIGGGKILAAALTECHRKSSAQGKPLALKVFVAGRNRLENDGATALAEAFRVIGTLEEVHMPQNGINHPGITALAQAFAVNPLLRVINLNDNTFTEKGAVAMAETLKTLRQVEVINFGDCLVRSKGAVAIADAIRGGLPKLKELNLSFCEIKRDAALAVAEAMADKAELEKLDLNGNTLGEEGCEQLQEVLEGFNMAKVLASLSDDEDEEEEEEGEEEEEEAEEEEEEDEEEEEEEEEEEEEEPQQRGQGEKSATPSRKILDPNTGEPAPVLSSPPPADVSTFLAFPSPEKLLRLGPKSSVLIAQQTDTSDPEKVVSAFLKVSSVFKDEATVRMAVQDAVDALMQKAFNSSSFNSNTFLTRLLVHMGLLKSEDKVKAIANLYGPLMALNHMVQQDYFPKALAPLLLAFVTKPNSALESCSFARHSLLQTLYKVSpecifica
AAH14044 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
90.31kDa | |
Glutathione Sepharose 4 Fast Flow | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
Fug1/KIAA1835/MGC20266/SD | |
RANGAP1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
5905 | |
RANGAP1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MASEDIAKLAETLAKTQVAGGQLSFKGKSLKLNTAEDAKDVIKEIEDFDSLEALRLEGNTVGVEAARVIAKALEKKSELKRCHWSDMFTGRLRTEIPPALISLGEGLITAGAQLVELDLSDNAFGPDGVQGFEALLKSSACFTLQELKLNNCGMGIGGGKILAAALTECHRKSSAQGKPLALKVFVAGRNRLENDGATALAEAFRVIGTLEEVHMPQNGINHPGITALAQAFAVNPLLRVINLNDNTFTEKGAVAMAETLKTLRQVEVINFGDCLVRSKGAVAIADAIRGGLPKLKELNLSFCEIKRDAALAVAEAMADKAELEKLDLNGNTLGEEGCEQLQEVLEGFNMAKVLASLSDDEDEEEEEEGEEEEEEAEEEEEEDEEEEEEEEEEEEEEPQQRGQGEKSATPSRKILDPNTGEPAPVLSSPPPADVSTFLAFPSPEKLLRLGPKSSVLIAQQTDTSDPEKVVSAFLKVSSVFKDEATVRMAVQDAVDALMQKAFNSSSFNSNTFLTRLLVHMGLLKSEDKVKAIANLYGPLMALNHMVQQDYFPKALAPLLLAFVTKPNSALESCSFARHSLLQTLYKV | |
RUO | |
RANGAP1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |