Learn More
Abnova™ Human RFT1 Partial ORF (NP_443091, 211 a.a. - 320 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00091869-Q01.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes an enzyme which catalyzes the translocation of the Man(5)GlcNAc (2)-PP-Dol intermediate from the cytoplasmic to the luminal side of the endoplasmic reticulum membrane in the pathway for the N-glycosylation of proteins. Mutations in this gene are associated with congenital disorder of glycosylation type In
Sequence: TKLQTLPVSRITDLLPNITRNGAFINWKEAKLTWSFFKQSFLKQILTEGERYVMTFLNVLNFGDQGVYDIVNNLGSLVARLIFQPIEESFYIFFAKVLERGKDATLQKQESpecifica
NP_443091 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.84kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
TKLQTLPVSRITDLLPNITRNGAFINWKEAKLTWSFFKQSFLKQILTEGERYVMTFLNVLNFGDQGVYDIVNNLGSLVARLIFQPIEESFYIFFAKVLERGKDATLQKQE | |
RUO | |
RFT1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
91869 | |
RFT1 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
CDG1N/DKFZp667J092/FLJ25945 | |
RFT1 | |
Recombinant | |
wheat germ expression system |