missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human ROPN1L Full-length ORF (AAH14607.1, 1 a.a. - 230 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00083853-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16125493

  • 349.00€ / 10µg
Spedizione stimata: 03-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

The protein encoded by this gene is a sperm protein, which interacts with A-kinase anchoring protein, AKAP3, through the amphipathic helix region of AKAP3. Type II regulatory subunit of cAMP-dependent protein kinase (PKARII) also binds to this helix domain of AKAP3, which allows PKARII to be targeted to specific subcellular compartments. It is suggested that sperm contains several proteins that bind to AKAPs in a manner similar to PKARII, and this encoded protein may be one of them. [provided by RefSeq]

Sequence: MPLPDTMFCAQQIHIPPELPDILKQFTKAAIRTQPADVLRWSAGYFSALSRGDPLPVKDRMEMPTATQKTDTGLTQGLLKVLHKQCHHKRYVELTDLEQKWKNLCLPKEKFKALLQLDPCENKIKWINFLALGCSMLGGSLNTALKHLCEILTDDREGGPARIPFKTFSYVYRYLARLDSDVSPLETESYLASLKENIDARKNGMIGLSDFFFPKRKLLESIENSEDVGH
Specifica

Specifica

AAH14607.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
52.6kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
ASP/FLJ23003/FLJ25776
ROPN1L
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
83853
ROPN1L (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MPLPDTMFCAQQIHIPPELPDILKQFTKAAIRTQPADVLRWSAGYFSALSRGDPLPVKDRMEMPTATQKTDTGLTQGLLKVLHKQCHHKRYVELTDLEQKWKNLCLPKEKFKALLQLDPCENKIKWINFLALGCSMLGGSLNTALKHLCEILTDDREGGPARIPFKTFSYVYRYLARLDSDVSPLETESYLASLKENIDARKNGMIGLSDFFFPKRKLLESIENSEDVGH
RUO
ROPN1L
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali