Learn More
Abcam Human RRBP1 knockout HeLa cell line
Codice prodotto. 32139894
Cambia vista
Click to view available options
2x1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32139894 2x1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32139894 Fornitore Abcam N. del fornitore ab2652462x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 13-10-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
RRBP1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 40 bp deletion in exon 3 and 41 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To order only knocko...
RRBP1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 40 bp deletion in exon 3 and 41 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells