Descrizione
RXRA KO cell line available to order. KO validated by Immunocytochemistry, Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, Homozygous: 1 bp insertion in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Specifications
| Quantità | 1,000,000 Cells |
| Status giuridico | RUO |