Learn More
Abcam Human S100A4 knockout A549 cell line
Codice prodotto. 32142227 Sfoglia Tutto Abcam Prodotti
Codice prodotto. 32142227 Fornitore Abcam N. del fornitore ab2618652x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 27-10-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
S100A4 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 1 bp insertion Frameshift = 95%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. ...
S100A4 KO cell line available to order. KO validated by Next Generation Sequencing, Western blot. Free of charge wild type control available. Knockout achieved by CRISPR/Cas9 X = 1 bp insertion Frameshift = 95%. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement