Learn More
Abcam Human SETD2 (KMT3A / HYPB / HIF-1) knockout HeLa cell line
Codice prodotto. 32157443 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
1,000,000 Cells
2 x 1,000,000 Cells
Dimensione della confezione:
Pezzo
pezzi 2
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32157443 2 x 1,000,000 Cells pezzi 2
32139793 1,000,000 Cells Pezzo
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Codice prodotto. 32157443 Fornitore Abcam N. del fornitore ab2617922x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 27-11-2026
SETD2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 3 and 2 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial. To or...
SETD2 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 3 and 2 bp deletion in exon 3. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells
Status giuridico RUO