Learn More
R&D Systems™ Human SLC31A1 (NP_001850.1) VersaClone cDNA
Specifica
Specifica
| Gene | SLC31A1/CTR1 |
| Numero di accesso | NP_001850.1 |
| ID gene (immissione) | 1317 |
| Alias gene | Copper transporter 1, COPT1, COPT1high affinity copper uptake protein 1, CTR1, CTR1MGC75487, HCTR1, hCTR1copper transport 1 homolog, solute carrier family 31 (copper transporters), member 1, Solute carrier family 31 member 1 |
| Content And Storage | Store the unopened product at -20 to -70 °C. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. |
| Specie | Human |
| Quantità | 10 μg |