missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC39A11 Full-length ORF (NP_631916.1, 1 a.a. - 335 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00201266-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16159273

  • 302.00€ / 10µg


Spedizione stimata
24-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MLQGHSSVFQALLGTFFTWGMTAAGAALVFVFSSGQRRILDGSLGFAAGVMLAASYWSLLAPAVEMATSSGGFGAFAFFPVAVGFTLGAAFVYLADLLMPHLGAAEDPQTALALNFGSTLMKKKSDPEGPALLFPESELSIRIDKSENGEAYQRKKAAATGLPEGPAVPVPSRGNLAQPGGSSWRRIALLILAITIHNVPEGLAVGVGFGAIEKTASATFESARNLAIGIGIQNFPEGLAVSLPLRGAGFSTWRAFWYGQLSGMVEPLAGVFGAFAVVLAEPILPYALAFAAGAMVYVVMDDIIPEAQISGNGKLASWASILGFVVMMSLDVGLG
Specifica

Specifica

NP_631916.1
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
61.1kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C17orf26
SLC39A11
Yes
wheat germ expression system
Liquid
201266
SLC39A11 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MLQGHSSVFQALLGTFFTWGMTAAGAALVFVFSSGQRRILDGSLGFAAGVMLAASYWSLLAPAVEMATSSGGFGAFAFFPVAVGFTLGAAFVYLADLLMPHLGAAEDPQTALALNFGSTLMKKKSDPEGPALLFPESELSIRIDKSENGEAYQRKKAAATGLPEGPAVPVPSRGNLAQPGGSSWRRIALLILAITIHNVPEGLAVGVGFGAIEKTASATFESARNLAIGIGIQNFPEGLAVSLPLRGAGFSTWRAFWYGQLSGMVEPLAGVFGAFAVVLAEPILPYALAFAAGAMVYVVMDDIIPEAQISGNGKLASWASILGFVVMMSLDVGLG
RUO
SLC39A11
Wheat Germ (in vitro)
GST
Videos
Documenti