missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SLC39A8 Full-length ORF (NP_071437.3, 1 a.a. - 460 a.a.) Recombinant Protein with GST-tag at N-terminal

Codice prodotto. p-7101777
Click to view available options
Quantità:
10 μg
25 μg
Dimensione della confezione:
10µg
25µg
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

Used for AP, Array, ELISA, WB-Re

This gene encodes a member of the SLC39 family of solute-carrier genes, which show structural characteristics of zinc transporters. The encoded protein is glycosylated and found in the plasma membrane and mitochondria, and functions in the cellular import of zinc at the onset of inflammation. It is also thought to be the primary transporter of the toxic cation cadmium, which is found in cigarette smoke. Multiple transcript variants encoding different isoforms have been found for this gene. Additional alternatively spliced transcript variants of this gene have been described, but their full-length nature is not known. [provided by RefSeq]

Sequence: MAPGRAVAGLLLLAAAGLGGVAEGPGLAFSEDVLSVFGANLSLSAAQLQHLLEQMGAASRVGVPEPGQLHFNQCLTAEEIFSLHGFSNATQITSSKFSVICPAVLQQLNFHPCEDRPKHKTRPSHSEVWGYGFLSVTIINLASLLGLILTPLIKKSYFPKILTFFVGLAIGTLFSNAIFQLIPEAFGFDPKVDSYVEKAVAVFGGFYLLFFFERMLKMLLKTYGQNGHTHFGNDNFGPQEKTHQPKALPAINGVTCYANPAVTEANGHIHFDNVSVVSLQDGKKEPSSCTCLKGPKLSEIGTIAWMITLCDALHNFIDGLAIGASCTLSLLQGLSTSIAILCEEFPHELGDFVILLNAGMSTRQALLFNFLSACSCYVGLAFGILVGNNFAPNIIFALAGGMFLYISLADMFPEMNDMLREKVTGRKTDFTFFMIQNAGMLTGFTAILLITLYAGEIELE

Specifications

Numero di accesso NP_071437.3
Da utilizzare con (applicazione) Antibody Production, Protein Array, ELISA, Western Blot
Formulazione 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID gene (immissione) 64116
Peso molecolare 76kDa
Nome SLC39A8 (Human) Recombinant Protein (P01)
Metodo di purificazione Glutathione Sepharose 4 Fast Flow
Analisi di controllo qualità 12.5% SDS-PAGE Stained with Coomassie Blue.
Quantità 10 μg
Immunogeno MAPGRAVAGLLLLAAAGLGGVAEGPGLAFSEDVLSVFGANLSLSAAQLQHLLEQMGAASRVGVPEPGQLHFNQCLTAEEIFSLHGFSNATQITSSKFSVICPAVLQQLNFHPCEDRPKHKTRPSHSEVWGYGFLSVTIINLASLLGLILTPLIKKSYFPKILTFFVGLAIGTLFSNAIFQLIPEAFGFDPKVDSYVEKAVAVFGGFYLLFFFERMLKMLLKTYGQNGHTHFGNDNFGPQEKTHQPKALPAINGVTCYANPAVTEANGHIHFDNVSVVSLQDGKKEPSSCTCLKGPKLSEIGTIAWMITLCDALHNFIDGLAIGASCTLSLLQGLSTSIAILCEEFPHELGDFVILLNAGMSTRQALLFNFLSACSCYVGLAFGILVGNNFAPNIIFALAGGMFLYISLADMFPEMNDMLREKVTGRKTDFTFFMIQNAGMLTGFTAILLITLYAGEIELE
Requisiti di stoccaggio Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Status giuridico RUO
Alias gene BIGM103/LZT-Hs6/PP3105/ZIP8
Nome comune SLC39A8
Simbolo del gene SLC39A8
Specie Wheat Germ (in vitro)
Recombinant Recombinant
Tag proteine GST
Sistema di espressione wheat germ expression system
Forma Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.