Learn More
Abnova™ Human SMO Partial ORF (NP_005622.1, 653 a.a. - 787 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca:  Abnova™ H00006608-Q02.25ug
Dettagli aggiuntivi : Peso : 0.00010kg
Codice prodotto. 16125535
  • 508.00€ / 25µg
Spedizione stimata: 28-08-2024
per visualizzare lo stock disponibile
Descrizione
Descrizione
Sequence: NLWLVEAEISPELQKRLGRKKKRRKRKKEVCPLAPPPELHPPAPAPSTIPRLPQLPRQKCLVAAGAWGAGDSCRQGAWTLVSNPFCPEPSPPQDPFLPSAPAPVAWAHGRRQGLGPIHSRTNLMDTELMDADSDF
Specifica
Specifica
NP_005622.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
40.37kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
NLWLVEAEISPELQKRLGRKKKRRKRKKEVCPLAPPPELHPPAPAPSTIPRLPQLPRQKCLVAAGAWGAGDSCRQGAWTLVSNPFCPEPSPPQDPFLPSAPAPVAWAHGRRQGLGPIHSRTNLMDTELMDADSDF
RUO
SMO
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
6608
SMO (Human) Recombinant Protein (Q02)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gx/SMOH
SMO
Recombinant
wheat germ expression system