missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human SPPL2A Partial ORF (NP_116191, 31 a.a. - 129 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00084888-Q01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16160927

  • 458.00€ / 25µg


Spedizione stimata
25-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: HASGNGTTKDYCMLYNPYWTALPSTLENATSISLMNLTSTPLCNLSDIPPVGIKSKAVVVPWGSCHFLEKARIAQKGGAEAMLVVNNSVLFPPSGNRSE
Specifica

Specifica

NP_116191
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.63kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
HASGNGTTKDYCMLYNPYWTALPSTLENATSISLMNLTSTPLCNLSDIPPVGIKSKAVVVPWGSCHFLEKARIAQKGGAEAMLVVNNSVLFPPSGNRSE
RUO
SPPL2A
Wheat Germ (in vitro)
GST
wheat germ expression system
Liquid
84888
SPPL2A (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
IMP3|PSL2
SPPL2A
Yes
Videos
Documenti