missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human TASP1 Full-length ORF (AAH25266, 1 a.a. - 420 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00055617-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16171603

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes an endopeptidase that cleaves specific substrates following aspartate residues. The encoded protein undergoes posttranslational autoproteolytic processing to generate alpha and beta subunits, which reassemble into the active alpha2-beta2 heterotetramer. It is required to cleave MLL, a protein required for the maintenance of HOX gene expression, and TFIIA, a basal transcription factor. Alternatively spliced transcript variants have been described, but their biological validity has not been determined. [provided by RefSeq]

Sequence: MTMEKGMSSGEGLPSRSSQVSAGKITAKELETKQSYKEKRGGFVLVHAGAGYHSESKAKEYKHVCKRACQKAIEKLQAGALATDAVTAALVELEDSPFTNAGMGSNLNLLGEIECDASIMDGKSLNFGAVGALSGIKNPVSVANRLLCEGQKGKLSAGRIPPCFLVGEGAYRWAVDHGIPSCPPNIMTTRFSLAAFKRNKRKLELAERVDTDFMQLKKRRQSSEKENDSGTLDTVGAVVVDHEGNVAAAVSSGGLALKHPGRVGQAALYGCGCWAENTGAHNPYSTAVSTSGCGEHLVRTILARECSHALQAEDAHQALLETMQNKFISSPFLASEDGVLGGVIVLRSCRCSAEPDSSQNKQTLLVEFLWSHTTESMCVGYMSAQDGKAKTHISRLPPGAVAGQSVAIEGGVCRLESPVN
Specifica

Specifica

AAH25266
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.94kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
C20orf13/FLJ20212/MGC39159/dJ585I14.2
TASP1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
55617
TASP1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTMEKGMSSGEGLPSRSSQVSAGKITAKELETKQSYKEKRGGFVLVHAGAGYHSESKAKEYKHVCKRACQKAIEKLQAGALATDAVTAALVELEDSPFTNAGMGSNLNLLGEIECDASIMDGKSLNFGAVGALSGIKNPVSVANRLLCEGQKGKLSAGRIPPCFLVGEGAYRWAVDHGIPSCPPNIMTTRFSLAAFKRNKRKLELAERVDTDFMQLKKRRQSSEKENDSGTLDTVGAVVVDHEGNVAAAVSSGGLALKHPGRVGQAALYGCGCWAENTGAHNPYSTAVSTSGCGEHLVRTILARECSHALQAEDAHQALLETMQNKFISSPFLASEDGVLGGVIVLRSCRCSAEPDSSQNKQTLLVEFLWSHTTESMCVGYMSAQDGKAKTHISRLPPGAVAGQSVAIEGGVCRLESPVN
RUO
TASP1
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali