Learn More
Abnova™ Human TCEAL1 Full-length ORF (AAH00809, 1 a.a. - 159 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifica
Numero di accesso | AAH00809 |
---|---|
Da utilizzare con (applicazione) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulazione | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID gene (immissione) | 9338 |
Peso molecolare | 43.23kDa |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16163542
|
Abnova™
H00009338-P01.10UG |
10 ug |
335.00€
10µg |
Spedizione stimata: 31-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16173542
|
Abnova™
H00009338-P01.25UG |
25 ug |
508.00€
25µg |
Spedizione stimata: 31-05-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
This gene encodes a member of the transcription elongation factor A (SII)-like (TCEAL) gene family. Members of this family may function as nuclear phosphoproteins that modulate transcription in a promoter context-dependent manner. The encoded protein is similar to transcription elongation factor A/transcription factor SII and contains a zinc finger-like motif as well as a sequence related to the transcription factor SII Pol II-binding region. It may exert its effects via protein-protein interactions with other transcriptional regulators rather than via direct binding of DNA. Multiple family members are located on the X chromosome. Alternative splicing results in multiple transcript variants encoding a single isoform. [provided by RefSeq]
Sequence: MDKPRKENEEEPQSAPKTDEERPPVEHSPEKQSPEEQSSEEQSSEEEFFPEELLPELLPEMLLSEERPPQEGLSRKDLFEGRPPMEQPPCGVGKHKLEEGSFKERLARSRPQFRGDIHGRNLSNEEMIQAADELEEMKRVRNKLMIMHWKAKRSRPYPISpecifica
AAH00809 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
43.23kDa | |
Glutathione Sepharose 4 Fast Flow | |
MDKPRKENEEEPQSAPKTDEERPPVEHSPEKQSPEEQSSEEQSSEEEFFPEELLPELLPEMLLSEERPPQEGLSRKDLFEGRPPMEQPPCGVGKHKLEEGSFKERLARSRPQFRGDIHGRNLSNEEMIQAADELEEMKRVRNKLMIMHWKAKRSRPYPI | |
RUO | |
TCEAL1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
9338 | |
TCEAL1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
SIIR/p21/pp21 | |
TCEAL1 | |
Recombinant | |
wheat germ expression system |