missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Human TGF beta-3 ELISA Kit

Codice prodotto. 31370017
Cambia vista
Click to view available options
Quantità:
96 Tests
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità
31370017 96 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso
Codice prodotto. 31370017 Fornitore Invitrogen™ N. del fornitore EH613RB
 Articolo soggetto a restrizioni
Spedizione stimata: 12-11-2026
Aggiungi al carrello
Aggiungi al carrello

missing translation for 'validMainframeMsg'

Les retours ne sont pas autorisés pour ce produit. Consulta la politica di reso

ELISA

Human TGF beta-3 ELISA Kit is an enzyme-linked immunosorbent assay for quantitative detection of human TGF beta 3 in cell culture supernatants and serum samples.

Transforming growth factor beta 3 (TGF beta 3) is the third member of the transforming growth factor family of cytokines, which also includes TGF beta 1 and beta 2. These cytokines are secreted in precursor form consisting of a bioactive C-terminal domain attached to an N-terminal domain known as latency associated protein (LAP). Cleavage of LAP results in the mature protein, which functions as a disulfide-linked homodimer. As with all members of the family, TGF beta 3 is highly conserved across species, with mouse and human TGF beta 3 demonstrating 100% sequence homology and cross-species activity.The members of this family can be expressed by most cell types and exert pleiotropic effects, which include the suppression of B- and T-cell effector activity, mediation of tissue healing, and suppression of tumor proliferation. The promotion of CD4+CD25+ T-cell expansion is a newly discovered function of the TGF beta cytokines, and indicates an important role in the protection against autoimmunity.
TRUSTED_SUSTAINABILITY

Specifica

Numero di accesso P10600
Intervallo saggio 4.3-1500 pg/mL
Assay Sensitivity 4.3 pg/mL
Coniugato HRP
Immunoassay Kit Format Sandwich ELISA Kit
Product Type ELISA
Tipo di campione Plasma, Serum, Supernatant
Target Species Human
Da utilizzare con (apparecchiatura) Absorbance Microplate Reader
Alias gene ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3ARVD; ARVD1; FLJ16571; H-TGF-b-3; LAP; Latency-associated peptide; LDS5; prepro-transforming growth factor beta-3; RNHF; TGF beta 3; Tgfb3; TGF-B3; Tgfb-3; TGF-beta3; TGF-beta-3; Transforming growth factor; transforming growth factor beta 3; transforming growth factor beta precursor; transforming growth factor beta-3; Transforming growth factor beta-3 proprotein; transforming growth factor, beta 3; transforming growth factor-beta3;
ID gene (immissione) 7043
Simbolo del gene TGFB3
Interassay CV 12%
Intraassay CV 10%
Contenuto kit Pre-coated microwell plate, Standard, Biotinylated Detection Antibody, Streptavidin-HRP, Sample Diluent, Wash Buffer Concentrate, Assay Buffer concentrate, Stop Solution, Substrate Solution, Adhesive Plate Covers
Label or Dye Biotin
Quantità 96 Tests
Status giuridico RUO
Shipping Condition Wet Ice
Requisiti di stoccaggio -20°C
Bersaglio TGF beta-3
Durata del test 1 hr 20 min
Tempo totale saggio 4 hr 45 min
Vedi altri risultati Mostra meno risultati
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.