Learn More
Abcam Human TPM4 knockout HEK-293T cell line
Codice prodotto. 32141485
Cambia vista
Click to view available options
2x1000000 Cells
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto.
32141485 2x1000000 Cells
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32141485 Fornitore Abcam N. del fornitore ab2666062x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 20-10-2026
Aggiungi al carrello
Læg i indkøbskurv
missing translation for 'validMainframeMsg'
Denne vare kan ikke returneres. Se returpolicy
TPM4 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 1 and Insertion of the selection cassette in exon 1. To order both knockout and wild-type control cells: select 2 x 10000...
TPM4 KO cell line available to order. KO validated by Western blot. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp insertion in exon 1 and Insertion of the selection cassette in exon 1. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.