Learn More
Abcam Human TRPM7 knockout HeLa cell line
Codice prodotto. 32143031 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
2 x 1,000,000 Cells
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32143031 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32143031 Fornitore Abcam N. del fornitore ab2654802x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 09-11-2026
TRPM7 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 14 and 5 bp deletion in exon 14 and 8 bp deletion in exon 14. To order both knockout and wild-type control cells: select 2 x 1000000 Cell...
TRPM7 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 1 bp deletion in exon 14 and 5 bp deletion in exon 14 and 8 bp deletion in exon 14. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifica
Quantità 2 x 1,000,000 Cells
Status giuridico RUO