Learn More
Abnova™ Human TTYH1 Full-length ORF (AAH19358.1, 1 a.a. - 239 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00057348-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a member of the tweety family of proteins. Members of this family function as chloride anion channels. The encoded protein functions as a calcium(2+)-independent, volume-sensitive large conductance chloride(-) channel. Two transcript variants encoding distinct isoforms have been identified for this gene. [provided by RefSeq]
Sequence: MWLAYVLLLLLELLVCLFTLLGLAKQSKWLVIVMTVMSLLVLVLSWGSMGLEAATAVGLSDFCSNPDPYVLNLTQEETGLSSDILSYYLLCNRAVSNPFQQRLTLSQRALANIHSQLLGLEREAVPQFPSAQKPLLSLEETLNVTEGNFHQLVALLHCRSLHKDYGAALRGLCEDALEGLLFLLLFSLLSAGALATALCSLPRAWALFPPSDDYDDTDDDDPFNPQQESKRFVQWQSSISpecifica
AAH19358.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
52.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
TTYH1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
57348 | |
TTYH1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MWLAYVLLLLLELLVCLFTLLGLAKQSKWLVIVMTVMSLLVLVLSWGSMGLEAATAVGLSDFCSNPDPYVLNLTQEETGLSSDILSYYLLCNRAVSNPFQQRLTLSQRALANIHSQLLGLEREAVPQFPSAQKPLLSLEETLNVTEGNFHQLVALLHCRSLHKDYGAALRGLCEDALEGLLFLLLFSLLSAGALATALCSLPRAWALFPPSDDYDDTDDDDPFNPQQESKRFVQWQSSI | |
RUO | |
TTYH1 | |
Recombinant | |
wheat germ expression system |