missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human UFC1 Full-length ORF (AAH05187.1, 1 a.a. - 167 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00051506-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
UFC1 is an E2-like conjugating enzyme for ubiquitin-fold modifier-1 (UFM1; MIM 610553) (Komatsu et al., 2004 [PubMed 15071506]).[supplied by OMIM]
Sequence: MADEATRRVVSEIPVLKTNAGPRDRELWVQRLKEEYQSLIRYVENNKNADNDWFRLESNKEGTRWFGKCWYIHDLLKYEFDIEFDIPITCPTTAPEIAVPELDGKTAKMYRGGKICLTDHFKPLWARNVPKFGLAHLMALGLGPWLAVEIPDLIQKGVIQHKEKCNQSpecifica
AAH05187.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
45.8kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSPC155 | |
UFC1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
51506 | |
UFC1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MADEATRRVVSEIPVLKTNAGPRDRELWVQRLKEEYQSLIRYVENNKNADNDWFRLESNKEGTRWFGKCWYIHDLLKYEFDIEFDIPITCPTTAPEIAVPELDGKTAKMYRGGKICLTDHFKPLWARNVPKFGLAHLMALGLGPWLAVEIPDLIQKGVIQHKEKCNQ | |
RUO | |
UFC1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |