Learn More
Abcam Human UFL1 knockout HEK-293T cell line
Codice prodotto. 32154433 Sfoglia Tutto Abcam Prodotti
Cambia vista
Click to view available options
Quantità:
2 x 1,000,000 Cells
Dimensione della confezione:
pezzi 2
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Codice prodotto. Quantità Dimensione della confezione
32154433 2 x 1,000,000 Cells pezzi 2
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Codice prodotto. 32154433 Fornitore Abcam N. del fornitore ab2665002x1000000Cells_
 Articolo soggetto a restrizioni
Spedizione stimata: 30-11-2026
Aggiungi al carrello
Aggiungi al carrello
missing translation for 'validMainframeMsg'
UFL1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 2 bp deletion in exon 2 and 5 bp deletion in exon 2 and 7 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vi...
UFL1 KO cell line available to order. KO validated. Free of charge wild type control available. Knockout achieved by using CRISPR/Cas9, 2 bp deletion in exon 2 and 5 bp deletion in exon 2 and 7 bp deletion in exon 2. To order both knockout and wild-type control cells: select 2 x 1000000 Cells/vial'. To order only knockout cells: select 1000000 Cells/vial'.
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Quantità 2 x 1,000,000 Cells
Status giuridico RUO
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.