missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human USP4 Full-length ORF (NP_003354.2, 1 a.a. - 963 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00007375-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16161762

  • 458.00€ / 25µg


Spedizione stimata
19-11-2020
Aggiungi al carrello

Descrizione

Descrizione

Sequence: MAEGGGCRERPDAETQKSELGPLMRTTLQRGAQWYLIDSRWFKQWKKYVGFDSWDMYNVGEHNLFPGPIDNSGLFSDPESQTLKEHLIDELDYVLVPTEAWNKLLNWYGCVEGQQPIVRKVVEHGLFVKHCKVEVYLLELKLCENSDPTNVLSCHFSKADTIATIEKEMRKLFNIPAERETRLWNKYMSNTYEQLSKLDNTVQDAGLYQGQVLVIEPQNEDGTWPRQTLQSKSSTAPSRNFTTSPKSSASPYSSVSASLIANGDSTSTCGMHSSGVSRGGSGFSASYNCQEPPSSHIQPGLCGLGNLGNTCFMNSALQCLSNTAPLTDYFLKDEYEAEINRDNPLGMKGEIAEAYAELIKQMWSGRDAHVAPRMFKTQVGRFAPQFSGYQQQDSQELLAFLLDGLHEDLNRVKKKPYLELKDANGRPDAVVAKEAWENHRLRNDSVIVDTFHGLFKSTLVCPECAKVSVTFDPFCYLTLPLPLKKDRVMEVFLVPADPHCRPTQYRVTVPLMGAVSDLCEALSRLSGIAAENMVVADVYNHRFHKIFQMDEGLNHIMPRDDIFVYEVCSTSVDGSECVTLPVYFRERKSRPSSTSSASALYGQPLLLSVPKHKLTLESLYQAVCDRISRYVKQPLPDEFGSSPLEPGACNGSRNSCEGEDEEEMEHQEEGKEQLSETEGSGEDEPGNDPSETTQKKIKGQPCPKRLFTFSLVNSYGTADINSLAADGKLLKLNSRSTLAMDWDSETRRLYYDEQESEAYEKHVSMLQPQKKKKTTVALRDCIELFTTMETLGEHDPWYCPNCKKHQQATKKFDLWSLPKILVVHLKRFSYNRYWRDKLDTVVEFPIRGLNMSEFVCNLSARPYVYDLIAVSNHYGAMGVGHYTAYAKNKLNGKWYYFDDSNVSLASEDQIVTKAAYVLFYQRRDDEFYKTPSLSSSGSSDGGTRPSSSQQGFGDDEACSMDTN
Specifica

Specifica

NP_003354.2
Antibody Production, Array, Enzyme-linked Immunoabsorbent Assay, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
135kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC149848|MGC149849|UNP|Unph
USP4
Yes
wheat germ expression system
Liquid
7375
USP4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEGGGCRERPDAETQKSELGPLMRTTLQRGAQWYLIDSRWFKQWKKYVGFDSWDMYNVGEHNLFPGPIDNSGLFSDPESQTLKEHLIDELDYVLVPTEAWNKLLNWYGCVEGQQPIVRKVVEHGLFVKHCKVEVYLLELKLCENSDPTNVLSCHFSKADTIATIEKEMRKLFNIPAERETRLWNKYMSNTYEQLSKLDNTVQDAGLYQGQVLVIEPQNEDGTWPRQTLQSKSSTAPSRNFTTSPKSSASPYSSVSASLIANGDSTSTCGMHSSGVSRGGSGFSASYNCQEPPSSHIQPGLCGLGNLGNTCFMNSALQCLSNTAPLTDYFLKDEYEAEINRDNPLGMKGEIAEAYAELIKQMWSGRDAHVAPRMFKTQVGRFAPQFSGYQQQDSQELLAFLLDGLHEDLNRVKKKPYLELKDANGRPDAVVAKEAWENHRLRNDSVIVDTFHGLFKSTLVCPECAKVSVTFDPFCYLTLPLPLKKDRVMEVFLVPADPHCRPTQYRVTVPLMGAVSDLCEALSRLSGIAAENMVVADVYNHRFHKIFQMDEGLNHIMPRDDIFVYEVCSTSVDGSECVTLPVYFRERKSRPSSTSSASALYGQPLLLSVPKHKLTLESLYQAVCDRISRYVKQPLPDEFGSSPLEPGACNGSRNSCEGEDEEEMEHQEEGKEQLSETEGSGEDEPGNDPSETTQKKIKGQPCPKRLFTFSLVNSYGTADINSLAADGKLLKLNSRSTLAMDWDSETRRLYYDEQESEAYEKHVSMLQPQKKKKTTVALRDCIELFTTMETLGEHDPWYCPNCKKHQQATKKFDLWSLPKILVVHLKRFSYNRYWRDKLDTVVEFPIRGLNMSEFVCNLSARPYVYDLIAVSNHYGAMGVGHYTAYAKNKLNGKWYYFDDSNVSLASEDQIVTKAAYVLFYQRRDDEFYKTPSLSSSGSSDGGTRPSSSQQGFGDDEACSMDTN
RUO
USP4
Wheat Germ (in vitro)
GST
Videos
Documenti