missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human USP4 Full-length ORF (NP_003354.2, 1 a.a. - 963 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifica
Numero di accesso | NP_003354.2 |
---|---|
Da utilizzare con (applicazione) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulazione | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
ID gene (immissione) | 7375 |
Peso molecolare | 135kDa |
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
---|---|---|---|---|---|---|---|---|---|
Codice del prodotto | Marca | Quantità | Prezzo | Quantità e disponibilità | |||||
16161762
|
Abnova™
H00007375-P01.25UG |
25 ug |
508.00€
25µg |
Spedizione stimata: 05-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
16151762
|
Abnova™
H00007375-P01.10UG |
10 ug |
335.00€
10µg |
Spedizione stimata: 05-06-2024 Eseguire il login per visualizzare lo stock disponibile |
Effettua il login per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso! | ||||
Descrizione
Sequence: MAEGGGCRERPDAETQKSELGPLMRTTLQRGAQWYLIDSRWFKQWKKYVGFDSWDMYNVGEHNLFPGPIDNSGLFSDPESQTLKEHLIDELDYVLVPTEAWNKLLNWYGCVEGQQPIVRKVVEHGLFVKHCKVEVYLLELKLCENSDPTNVLSCHFSKADTIATIEKEMRKLFNIPAERETRLWNKYMSNTYEQLSKLDNTVQDAGLYQGQVLVIEPQNEDGTWPRQTLQSKSSTAPSRNFTTSPKSSASPYSSVSASLIANGDSTSTCGMHSSGVSRGGSGFSASYNCQEPPSSHIQPGLCGLGNLGNTCFMNSALQCLSNTAPLTDYFLKDEYEAEINRDNPLGMKGEIAEAYAELIKQMWSGRDAHVAPRMFKTQVGRFAPQFSGYQQQDSQELLAFLLDGLHEDLNRVKKKPYLELKDANGRPDAVVAKEAWENHRLRNDSVIVDTFHGLFKSTLVCPECAKVSVTFDPFCYLTLPLPLKKDRVMEVFLVPADPHCRPTQYRVTVPLMGAVSDLCEALSRLSGIAAENMVVADVYNHRFHKIFQMDEGLNHIMPRDDIFVYEVCSTSVDGSECVTLPVYFRERKSRPSSTSSASALYGQPLLLSVPKHKLTLESLYQAVCDRISRYVKQPLPDEFGSSPLEPGACNGSRNSCEGEDEEEMEHQEEGKEQLSETEGSGEDEPGNDPSETTQKKIKGQPCPKRLFTFSLVNSYGTADINSLAADGKLLKLNSRSTLAMDWDSETRRLYYDEQESEAYEKHVSMLQPQKKKKTTVALRDCIELFTTMETLGEHDPWYCPNCKKHQQATKKFDLWSLPKILVVHLKRFSYNRYWRDKLDTVVEFPIRGLNMSEFVCNLSARPYVYDLIAVSNHYGAMGVGHYTAYAKNKLNGKWYYFDDSNVSLASEDQIVTKAAYVLFYQRRDDEFYKTPSLSSSGSSDGGTRPSSSQQGFGDDEACSMDTNSpecifica
NP_003354.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
135kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAEGGGCRERPDAETQKSELGPLMRTTLQRGAQWYLIDSRWFKQWKKYVGFDSWDMYNVGEHNLFPGPIDNSGLFSDPESQTLKEHLIDELDYVLVPTEAWNKLLNWYGCVEGQQPIVRKVVEHGLFVKHCKVEVYLLELKLCENSDPTNVLSCHFSKADTIATIEKEMRKLFNIPAERETRLWNKYMSNTYEQLSKLDNTVQDAGLYQGQVLVIEPQNEDGTWPRQTLQSKSSTAPSRNFTTSPKSSASPYSSVSASLIANGDSTSTCGMHSSGVSRGGSGFSASYNCQEPPSSHIQPGLCGLGNLGNTCFMNSALQCLSNTAPLTDYFLKDEYEAEINRDNPLGMKGEIAEAYAELIKQMWSGRDAHVAPRMFKTQVGRFAPQFSGYQQQDSQELLAFLLDGLHEDLNRVKKKPYLELKDANGRPDAVVAKEAWENHRLRNDSVIVDTFHGLFKSTLVCPECAKVSVTFDPFCYLTLPLPLKKDRVMEVFLVPADPHCRPTQYRVTVPLMGAVSDLCEALSRLSGIAAENMVVADVYNHRFHKIFQMDEGLNHIMPRDDIFVYEVCSTSVDGSECVTLPVYFRERKSRPSSTSSASALYGQPLLLSVPKHKLTLESLYQAVCDRISRYVKQPLPDEFGSSPLEPGACNGSRNSCEGEDEEEMEHQEEGKEQLSETEGSGEDEPGNDPSETTQKKIKGQPCPKRLFTFSLVNSYGTADINSLAADGKLLKLNSRSTLAMDWDSETRRLYYDEQESEAYEKHVSMLQPQKKKKTTVALRDCIELFTTMETLGEHDPWYCPNCKKHQQATKKFDLWSLPKILVVHLKRFSYNRYWRDKLDTVVEFPIRGLNMSEFVCNLSARPYVYDLIAVSNHYGAMGVGHYTAYAKNKLNGKWYYFDDSNVSLASEDQIVTKAAYVLFYQRRDDEFYKTPSLSSSGSSDGGTRPSSSQQGFGDDEACSMDTN | |
RUO | |
USP4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
7375 | |
USP4 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC149848/MGC149849/UNP/Unph | |
USP4 | |
Recombinant | |
wheat germ expression system |