missing translation for 'onlineSavingsMsg'
Learn More
Learn More
R&D Systems™ Human VEGF DuoSet ELISA
Descrizione
The Human VEGF DuoSet ELISA is a Solid Phase Sandwich ELISA that quantifies human VEGF.
Specifica
Specifica
| Product Type | ELISA Development Kit |
| Quantità | 1 Kit |
| Specie | Human |
| Coniugato | Biotin |
| Intervallo saggio | 31.20 to 2000 pg/mL |
| Cross-reattività | <50% cross-species reactivity observed with species tested. <0.5% cross-reactivity observed with available related molecules |
| Da utilizzare con (applicazione) | ELISA |
| ID gene (immissione) | 7422 |
| Quantità confezione | 15 Plates |
| Content And Storage | Store the unopened product at 2°C to 8°C. Do not use past expiration date. |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?