missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VEGFA Full-length ORF (ENSP00000361134, 1 a.a. - 147 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifica

Numero di accesso ENSP00000361134
Da utilizzare con (applicazione) Antibody Production, ELISA, Protein Array, Western Blot
Formulazione 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
ID gene (immissione) 7422
Peso molecolare 43.6kDa
Visualizza altre specifiche

Prodotti 2
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
Codice del prodotto Marca Quantità Prezzo Quantità e disponibilità  
16171822
Vedere i documenti Dettagli della promozione
Abnova™
H00007422-P01.25UG
25 ug
508.00€
25µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 10-05-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
16161822
Vedere i documenti Dettagli della promozione
Abnova™
H00007422-P01.10UG
10 ug
335.00€
10µg
Visualizzare prodotti alternativi
Risparmia fino a 
»
Spedizione stimata: 10-05-2024
per visualizzare lo stock disponibile
Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!
Descrizione

Descrizione

This gene is a member of the PDGF/VEGF growth factor family and encodes a protein that is often found as a disulfide linked homodimer. This protein is a glycosylated mitogen that specifically acts on endothelial cells and has various effects, including mediating increased vascular permeability, inducing angiogenesis, vasculogenesis and endothelial cell growth, promoting cell migration, and inhibiting apoptosis. Elevated levels of this protein is linked to POEMS syndrome, also known as Crow-Fukase syndrome. Mutations in this gene have been associated with proliferative and nonproliferative diabetic retinopathy. Alternatively spliced transcript variants, encoding either freely secreted or cell-associated isoforms, have been characterized. There is also evidence for the use of non-AUG (CUG) translation initiation sites upstream of, and in-frame with the first AUG, leading to additional isoforms. [provided by RefSeq]

Sequence: MNFLLSWVHWSLALLLYLHHAKWSQAAPMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGCCNDEGLECVPTEESNITMQIMRIKPHQGQHIGEMSFLQHNKCECRPKKDRARQEKCDKPRR
Specifica

Specifica

ENSP00000361134
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
43.6kDa
Glutathione Sepharose 4 Fast Flow
MNFLLSWVHWSLALLLYLHHAKWSQAAPMAEGGGQNHHEVVKFMDVYQRSYCHPIETLVDIFQEYPDEIEYIFKPSCVPLMRCGGCCNDEGLECVPTEESNITMQIMRIKPHQGQHIGEMSFLQHNKCECRPKKDRARQEKCDKPRR
RUO
VEGFA
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
7422
VEGFA (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC70609/VEGF/VEGF-A/VPF
VEGFA
Recombinant
wheat germ expression system
Videos
SDS
Documenti

Documenti

Product Certifications