missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human VNN2 Full-length ORF (NP_511043.1, 1 a.a. - 467 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00008875-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16133032

  • 349.00€ / 10µg
Spedizione stimata: 07-11-2023
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene product is a member of the Vanin family of proteins which share extensive sequence similarity with each other, and also with biotinidase. The family includes secreted and membrane-associated proteins, a few of which have been reported to participate in hematopoietic cell trafficking. No biotinidase activity has been demonstrated for any of the vanin proteins, however, they possess pantetheinase activity, which may play a role in oxidative-stress response. The encoded protein is a GPI-anchored cell surface molecule that plays a role in transendothelial migration of neutrophils. This gene lies in close proximity to, and in same transcriptional orientation as two other vanin genes on chromosome 6q23-q24. Two transcript variants encoding different isoforms have been described for this gene. [provided by RefSeq]

Sequence: MNENIDILETAIKQAAEQGARIIVTPEDALYGWKFTRETVFPYLEDIPDPQVNWIPCQDPHRFGHTPVQARLSCLAKDNSIYVLANLGDKKPCNSRDSTCPPNGYFQYNTNVVYNTEGKLVARYHKYHLYSEPQFNVPEKPELVTFNTAFGRFGIFTCFDIFFYDPGVTLVKDFHVDTILFPTAWMNVLPLLTAIEFHSAWAMGMGVNLLVANTHHVSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSCGTSNSAITYLLIFILLMIIALQNIVML
Specifica

Specifica

NP_511043.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
79.3kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FOAP-4/GPI-80
VNN2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
8875
VNN2 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNENIDILETAIKQAAEQGARIIVTPEDALYGWKFTRETVFPYLEDIPDPQVNWIPCQDPHRFGHTPVQARLSCLAKDNSIYVLANLGDKKPCNSRDSTCPPNGYFQYNTNVVYNTEGKLVARYHKYHLYSEPQFNVPEKPELVTFNTAFGRFGIFTCFDIFFYDPGVTLVKDFHVDTILFPTAWMNVLPLLTAIEFHSAWAMGMGVNLLVANTHHVSLNMTGSGIYAPNGPKVYHYDMKTELGKLLLSEVDSHPLSSLAYPTAVNWNAYATTIKPFPVQKNTFRGFISRDGFNFTELFENAGNLTVCQKELCCHLSYRMLQKEENEVYVLGAFTGLHGRRRREYWQVCTMLKCKTTNLTTCGRPVETASTRFEMFSLSGTFGTEYVFPEVLLTEIHLSPGKFEVLKDGRLVNKNGSSGPILTVSLFGRWYTKDSLYSSCGTSNSAITYLLIFILLMIIALQNIVML
RUO
VNN2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali