Learn More
Abnova™ Human VPS52 Full-length ORF (NP_072047.4, 1 a.a. - 723 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Marca: Abnova™ H00006293-P01.10ug
Dettagli aggiuntivi : Peso : 0.00010kg
Descrizione
This gene encodes a protein that is similar to the yeast suppressor of actin mutations 2 gene. The yeast protein forms a subunit of the tetrameric Golgi-associated retrograde protein complex that is involved in vesicle trafficking from from both early and late endosomes, back to the trans-Golgi network. This gene is located on chromosome 6 in a head-to-head orientation with the gene encoding ribosomal protein S18. [provided by RefSeq]
Sequence: MAAAATMAAAARELVLRAGTSDMEEEEGPLAGGPGLQEPLQLGELDITSDEFILDEVDVHIQANLEDELVKEALKTGVDLRHYSKQVELELQQIEQKSIRDYIQESENIASLHNQITACDAVLERMEQMLGAFQSDLSSISSEIRTLQEQSGAMNIRLRNRQAVRGKLGELVDGLVVPSALVTAILEAPVTEPRFLEQLQELDAKAAAVREQEARGTAACADVRGVLDRLRVKAVTKIREFILQKIYSFRKPMTNYQIPQTALLKYRFFYQFLLGNERATAKEIRDEYVETLSKIYLSYYRSYLGRLMKVQYEEVAEKDDLMGVEDTAKKGFFSKPSLRSRNTIFTLGTRGSVISPTELEAPILVPHTAQRGEQRYPFEALFRSQHYALLDNSCREYLFICEFFVVSGPAAHDLFHAVMGRTLSMTLKHLDSYLADCYDAIAVFLCIHIVLRFRNIAAKRDVPALDRYWEQVLALLWPRFELILEMNVQSVRSTDPQRLGGLDTRPHYITRRYAEFSSALVSINQTIPNERTMQLLGQLQVEVENFVLRVAAEFSSRKEQLVFLINNYDMMLGVLMERAADDSKEVESFQQLLNARTQEFIEELLSPPFGGLVAFVKEAEALIERGQAERLRGEEARVTQLIRGFGSSWKSSVESLSQDVMRSFTNFRNGTSIIQGALTQLIQLYHRFHRVLSQPQLRALPARAELINIHHLMVELKKHKPNFSpecifica
NP_072047.4 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
ARE1/DKFZp547I194/RP5-1033B10/SAC2/SACM2L/dJ1033B10.5 | |
VPS52 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
6293 | |
VPS52 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAAAATMAAAARELVLRAGTSDMEEEEGPLAGGPGLQEPLQLGELDITSDEFILDEVDVHIQANLEDELVKEALKTGVDLRHYSKQVELELQQIEQKSIRDYIQESENIASLHNQITACDAVLERMEQMLGAFQSDLSSISSEIRTLQEQSGAMNIRLRNRQAVRGKLGELVDGLVVPSALVTAILEAPVTEPRFLEQLQELDAKAAAVREQEARGTAACADVRGVLDRLRVKAVTKIREFILQKIYSFRKPMTNYQIPQTALLKYRFFYQFLLGNERATAKEIRDEYVETLSKIYLSYYRSYLGRLMKVQYEEVAEKDDLMGVEDTAKKGFFSKPSLRSRNTIFTLGTRGSVISPTELEAPILVPHTAQRGEQRYPFEALFRSQHYALLDNSCREYLFICEFFVVSGPAAHDLFHAVMGRTLSMTLKHLDSYLADCYDAIAVFLCIHIVLRFRNIAAKRDVPALDRYWEQVLALLWPRFELILEMNVQSVRSTDPQRLGGLDTRPHYITRRYAEFSSALVSINQTIPNERTMQLLGQLQVEVENFVLRVAAEFSSRKEQLVFLINNYDMMLGVLMERAADDSKEVESFQQLLNARTQEFIEELLSPPFGGLVAFVKEAEALIERGQAERLRGEEARVTQLIRGFGSSWKSSVESLSQDVMRSFTNFRNGTSIIQGALTQLIQLYHRFHRVLSQPQLRALPARAELINIHHLMVELKKHKPNF | |
RUO | |
VPS52 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |