missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WDR57 Full-length ORF (AAH01494.1, 1 a.a. - 357 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00009410-P01.10ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16163632

  • 335.00€ / 10µg
Spedizione stimata: 07-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This gene encodes a component of the U5 small nuclear ribonucleoprotein (snRNP) particle. The U5 snRNP is part of the spliceosome, a multiprotein complex that catalyzes the removal of introns from pre-messenger RNAs. [provided by RefSeq]

Sequence: MIEQQKRKGPELPLVPVKRQRHELLLGAGSGPGAGQQQATPGALLQAGPPRCSSLQAPIMLLSGHEGEVYCCKFHPNGSTLASAGFDRLILLWNVYGDCDNYATLKGHSGAVMELHYNTDGSMLFSASTDKTVAVWDSETGERVKRLKGHTSFVNSCYPARRGPQLVCTGSDDGTVKLWDIRKKAAIQTFQNTYQVLAVTFNDTSDQIISGGIDNDIKVWDLRQNKLTYTMRGHADSVTGLSLSSEGSYLLSNAMDNTVRVWDVRPFAPKERCVKIFQGNVHNFEKNLLRCSWSPDGSKIAAGSADRFVYVWDTTSRRILYKLPGHAGSINEVAFHPDEPIIISASSDKRLYMGEIQ
Specifica

Specifica

AAH01494.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
65.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
40K/FLJ41108/HPRP8BP/MGC1910/PRP8BP/PRPF8BP/RP11-490K7.3/SPF38/WDR57
SNRNP40
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
9410
WDR57 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MIEQQKRKGPELPLVPVKRQRHELLLGAGSGPGAGQQQATPGALLQAGPPRCSSLQAPIMLLSGHEGEVYCCKFHPNGSTLASAGFDRLILLWNVYGDCDNYATLKGHSGAVMELHYNTDGSMLFSASTDKTVAVWDSETGERVKRLKGHTSFVNSCYPARRGPQLVCTGSDDGTVKLWDIRKKAAIQTFQNTYQVLAVTFNDTSDQIISGGIDNDIKVWDLRQNKLTYTMRGHADSVTGLSLSSEGSYLLSNAMDNTVRVWDVRPFAPKERCVKIFQGNVHNFEKNLLRCSWSPDGSKIAAGSADRFVYVWDTTSRRILYKLPGHAGSINEVAFHPDEPIIISASSDKRLYMGEIQ
RUO
SNRNP40
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali