missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human WDR5B Full-length ORF (NP_061942.2, 1 a.a. - 330 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Marca:  Abnova™ H00054554-P01.25ug

Dettagli aggiuntivi : Peso : 0.00010kg

 vedi altre versioni di questo prodotto

Codice prodotto. 16160643

  • 508.00€ / 25µg
Spedizione stimata: 10-05-2024
per visualizzare lo stock disponibile

Effettua il per acquistare questo articolo Hai bisogno di un conto web ? Registrati oggi stesso!


Descrizione

Descrizione

This intronless gene encodes a protein containing several WD40 repeats. WD repeats are approximately 30- to 40-amino acid domains containing several conserved residues, including a trp-asp at the C-terminal end. The encoded protein may mediate protein-protein interactions. [provided by RefSeq]

Sequence: MATKESRDAKAQLALSSSANQSKEVPENPNYALKCTLVGHTEAVSSVKFSPNGEWLASSSADRLIIIWGAYDGKYEKTLYGHNLEISDVAWSSDSSRLVSASDDKTLKLWDVRSGKCLKTLKGHSNYVFCCNFNPPSNLIISGSFDETVKIWEVKTGKCLKTLSAHSDPVSAVHFNCSGSLIVSGSYDGLCRIWDAASGQCLKTLVDDDNPPVSFVKFSPNGKYILTATLDNTLKLWDYSRGRCLKTYTGHKNEKYCIFANFSVTGGKWIVSGSEDNLVYIWNLQTKEIVQKLQGHTDVVISAACHPTENLIASAALENDKTIKLWMSNH
Specifica

Specifica

NP_061942.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
62.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ11287/MGC49879
WDR5B
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
54554
WDR5B (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MATKESRDAKAQLALSSSANQSKEVPENPNYALKCTLVGHTEAVSSVKFSPNGEWLASSSADRLIIIWGAYDGKYEKTLYGHNLEISDVAWSSDSSRLVSASDDKTLKLWDVRSGKCLKTLKGHSNYVFCCNFNPPSNLIISGSFDETVKIWEVKTGKCLKTLSAHSDPVSAVHFNCSGSLIVSGSYDGLCRIWDAASGQCLKTLVDDDNPPVSFVKFSPNGKYILTATLDNTLKLWDYSRGRCLKTYTGHKNEKYCIFANFSVTGGKWIVSGSEDNLVYIWNLQTKEIVQKLQGHTDVVISAACHPTENLIASAALENDKTIKLWMSNH
RUO
WDR5B
Wheat Germ (in vitro)
GST
Liquid
Suggerimenti di prodotto

Suggerimenti di prodotto

Videos
SDS
Documenti

Documenti

Certificati
Offerte speciali

Offerte speciali